Assay Details
| Gene Symbol: usp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008574.2 | NP_001008574.2 | 5 - 6 | 831 | 140 | |
| XM_005157513.4 | XP_005157570.1 | 7 - 8 | 1331 | 140 | ||
| XM_005157514.4 | XP_005157571.1 | 7 - 8 | 1425 | 140 | ||
| XM_005157515.4 | XP_005157572.1 | 7 - 8 | 1326 | 140 | ||
| XM_005157516.4 | XP_005157573.1 | 5 - 6 | 899 | 140 | ||
| XM_017350951.2 | XP_017206440.1 | 7 - 8 | 1326 | 140 | ||
| XM_021481396.1 | XP_021337071.1 | 7 - 8 | 1429 | 140 | ||
| GenBank mRNA | BC086963.1 | - | 5 - 6 | 672 | 140 | |
More Information
Set Membership: |
|

usp2a
Genomic Map