Assay Details
| Gene Symbol: nabp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008643.1 | NP_001008643.1 | 4 - 5 | 604 | 88 | |
| GenBank mRNA | BC086713.1 | - | 4 - 5 | 604 | 88 | |
More Information
Set Membership: |
|

nabp1a
Genomic Map