Assay Details
| Gene Symbol: cdcp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014333.1 | NP_001014355.1 | 5 - 6 | 1274 | 82 | |
| GenBank mRNA | BC091838.1 | - | 5 - 6 | 1274 | 82 | |
| BC164573.1 | - | 5 - 6 | 1199 | 82 | ||
More Information
Set Membership: |
|

cdcp1a
Genomic Map