Assay Details
| Gene Symbol: cdcp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014333.1 | NP_001014355.1 | 8 - 9 | 2090 | 102 | |
| GenBank mRNA | BC091838.1 | - | 8 - 9 | 2090 | 102 | |
| BC164573.1 | - | 8 - 9 | 2015 | 102 | ||
More Information
Set Membership: |
|

cdcp1a
Genomic Map