Assay Details
| Gene Symbol: rbm39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014370.1 | NP_001014392.1 | 5 - 6 | 601 | 56 | |
| XM_005167222.4 | XP_005167279.1 | 4 - 5 | 551 | 56 | ||
| XM_005167224.4 | XP_005167281.1 | 5 - 6 | 626 | 56 | ||
| XM_009304133.3 | XP_009302408.1 | 6 - 7 | 772 | 56 | ||
| XM_021478249.1 | XP_021333924.1 | 4 - 5 | 564 | 56 | ||
| XM_021478250.1 | XP_021333925.1 | 4 - 5 | 452 | 56 | ||
| XM_021478251.1 | XP_021333926.1 | 4 - 5 | 551 | 56 | ||
| GenBank mRNA | BC091794.1 | - | 5 - 6 | 601 | 56 | |
| BC093217.1 | - | 4 - 5 | 532 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005167224.4, XM_009304133.3, XM_021478249.1, XM_021478250.1 transcripts.
Set Membership: |
|

rbm39b
Genomic Map