Assay Details
| Gene Symbol: cntn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014814.1 | NP_001014814.1 | 12 - 13 | 1619 | 88 | |
| GenBank mRNA | AY301281.1 | - | 12 - 13 | 1619 | 88 | |
More Information
Set Membership: |
|

cntn1b
Genomic Map