Assay Details
| Gene Symbol: exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017572.2 | NP_001017572.2 | 7 - 8 | 743 | 57 | |
| GenBank mRNA | BC092739.1 | - | 7 - 8 | 706 | 57 | |
| BC164830.1 | - | 6 - 7 | 588 | 57 | ||
| EH525603.1 | - | 2 - 3 | 287 | 57 | ||
| EH545647.1 | - | 7 - 8 | 765 | 57 | ||
More Information
Set Membership: |
|

exosc2
Genomic Map