Assay Details
| Gene Symbol: srd5a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017703.1 | NP_001017703.1 | 3 - 4 | 584 | 58 | |
| GenBank mRNA | BC076038.1 | - | 2 - 3 | 1170 | 58 | |
| BC093267.1 | - | 3 - 4 | 584 | 58 | ||
| BC095069.1 | - | 3 - 4 | 584 | 58 | ||
| BC164829.1 | - | 3 - 4 | 569 | 58 | ||
More Information
Set Membership: |
|

srd5a2a
Genomic Map