Assay Details
| Gene Symbol: grk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017711.2 | NP_001017711.2 | 2 - 3 | 926 | 95 | |
| XM_021476153.1 | XP_021331828.1 | 2 - 3 | 989 | 95 | ||
| GenBank mRNA | AB212994.1 | - | 2 - 3 | 926 | 95 | |
| AY900003.1 | - | 1 - 2 | 750 | 95 | ||
| BC093257.1 | - | 2 - 3 | 825 | 95 | ||
More Information
Set Membership: |
|

grk1b
Genomic Map