Assay Details
| Gene Symbol: grk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017711.2 | NP_001017711.2 | 3 - 4 | 1049 | 153 | |
| GenBank mRNA | AB212994.1 | - | 3 - 4 | 1049 | 153 | |
| AY900003.1 | - | 2 - 3 | 873 | 153 | ||
| BC093257.1 | - | 3 - 4 | 948 | 153 | ||
More Information
Set Membership: |
|

grk1b
Genomic Map