Assay Details
| Gene Symbol: gucy1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309529.1 | NP_001296458.1 | 4 - 5 | 1306 | 143 | |
| GenBank mRNA | BC093232.1 | - | 4 - 5 | 1301 | 143 | |
| HE577852.1 | - | 4 - 5 | 1102 | 143 | ||
More Information
Set Membership: |
|

gucy1a1
Genomic Map