Assay Details
| Gene Symbol: rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017740.1 | NP_001017740.1 | 2 - 3 | 204 | 135 | |
| GenBank mRNA | BC093208.1 | - | 2 - 3 | 204 | 135 | |
More Information
Set Membership: |
|

rfesd
Genomic Map