Assay Details
| Gene Symbol: syt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020546.2 | NP_001018382.2 | 7 - 8 | 1015 | 105 | |
| GenBank mRNA | BC095109.1 | - | 7 - 8 | 1015 | 105 | |
| BC165563.1 | - | 6 - 7 | 932 | 105 | ||
More Information
Set Membership: |
|

syt5b
Genomic Map