Assay Details
| Gene Symbol: acbd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020647.1 | NP_001018483.1 | 5 - 6 | 463 | 65 | |
| XM_005162509.4 | XP_005162566.1 | 5 - 6 | 485 | 65 | ||
| GenBank mRNA | BC095655.1 | - | 5 - 6 | 463 | 65 | |
More Information
Set Membership: |
|

acbd5a
Genomic Map