Assay Details
| Gene Symbol: ptp4a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024098.2 | NP_001019269.1 | 2 - 3 | 769 | 64 | |
| NM_001270540.1 | NP_001257469.1 | 3 - 4 | 490 | 64 | ||
| GenBank mRNA | AY394936.1 | - | 2 - 3 | 720 | 64 | |
| BC066747.1 | - | 1 - 2 | 351 | 64 | ||
| BC085424.1 | - | 3 - 4 | 470 | 64 | ||
| CT685667.2 | - | 1 - 2 | 409 | 64 | ||
| DT076629.1 | - | 1 - 2 | 45 | 64 | ||
More Information
Set Membership: |
|

ptp4a2b
Genomic Map