Assay Details
| Gene Symbol: ptp4a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024098.2 | NP_001019269.1 | 4 - 5 | 996 | 68 | |
| NM_001270540.1 | NP_001257469.1 | 5 - 6 | 717 | 68 | ||
| GenBank mRNA | AY394936.1 | - | 4 - 5 | 947 | 68 | |
| BC066747.1 | - | 3 - 4 | 578 | 68 | ||
| BC085424.1 | - | 5 - 6 | 697 | 68 | ||
| CT685667.2 | - | 3 - 4 | 636 | 68 | ||
| DT076629.1 | - | 3 - 4 | 272 | 68 | ||
More Information
Set Membership: |
|

ptp4a2b
Genomic Map