Assay Details
| Gene Symbol: ptp4a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024098.2 | NP_001019269.1 | 5 - 6 | 1076 | 90 | |
| NM_001270540.1 | NP_001257469.1 | 6 - 7 | 797 | 90 | ||
| GenBank mRNA | AY394936.1 | - | 5 - 6 | 1027 | 90 | |
| BC085424.1 | - | 6 - 7 | 777 | 90 | ||
| CT685667.2 | - | 4 - 5 | 716 | 90 | ||
| DT076629.1 | - | 4 - 5 | 352 | 90 | ||
More Information
Set Membership: |
|

ptp4a2b
Genomic Map