Assay Details
| Gene Symbol: slc27a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025299.1 | NP_001020470.1 | 3 - 4 | 839 | 74 | |
| GenBank mRNA | BC068405.1 | - | 3 - 4 | 819 | 74 | |
| BC098625.1 | - | 3 - 4 | 839 | 74 | ||
More Information
Set Membership: |
|

slc27a2a
Genomic Map