Assay Details
| Gene Symbol: slc27a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025299.1 | NP_001020470.1 | 4 - 5 | 965 | 101 | |
| GenBank mRNA | BC068405.1 | - | 4 - 5 | 945 | 101 | |
| BC098625.1 | - | 4 - 5 | 965 | 101 | ||
More Information
Set Membership: |
|

slc27a2a
Genomic Map