Assay Details
| Gene Symbol: pak2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025456.1 | NP_001020627.1 | 11 - 12 | 1405 | 77 | |
| XM_005170360.4 | XP_005170417.1 | 11 - 12 | 77 | |||
| GenBank mRNA | BC096967.1 | - | 11 - 12 | 1405 | 77 | |
More Information
Set Membership: |
|

pak2b
Genomic Map