Assay Details
| Gene Symbol: pak2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025456.1 | NP_001020627.1 | 13 - 14 | 1701 | 65 | |
| XM_005170360.4 | XP_005170417.1 | 13 - 14 | 1572 | 65 | ||
| GenBank mRNA | BC096967.1 | - | 13 - 14 | 1701 | 65 | |
More Information
Set Membership: |
|

pak2b
Genomic Map