Assay Details
| Gene Symbol: pak2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025456.1 | NP_001020627.1 | 5 - 6 | 779 | 90 | |
| XM_005170360.4 | XP_005170417.1 | 5 - 6 | 656 | 90 | ||
| GenBank mRNA | BC081414.1 | - | 5 - 6 | 636 | 90 | |
| BC095627.1 | - | 5 - 6 | 750 | 90 | ||
| BC096967.1 | - | 5 - 6 | 779 | 90 | ||
More Information
Set Membership: |
|

pak2b
Genomic Map