Assay Details
| Gene Symbol: rtn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029965.3 | NP_001025136.1 | 5 - 6 | 543 | 75 | |
| XM_005157471.4 | XP_005157528.1 | 8 - 9 | 1352 | 75 | ||
| XM_005157472.4 | XP_005157529.1 | 5 - 6 | 591 | 75 | ||
| GenBank mRNA | AY576807.1 | - | 5 - 6 | 543 | 75 | |
| BC115054.1 | - | 5 - 6 | 541 | 75 | ||
More Information
Set Membership: |
|

rtn2a
Genomic Map