Assay Details
| Gene Symbol: srgap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030095.1 | NP_001025266.1 | 11 - 12 | 1955 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srgap1b| Gene Symbol: srgap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030095.1 | NP_001025266.1 | 11 - 12 | 1955 | 127 | |
Set Membership: |
|