Assay Details
| Gene Symbol: srgap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030095.1 | NP_001025266.1 | 1 - 2 | 539 | 136 | |
| XM_005164583.4 | XP_005164640.1 | 1 - 2 | 557 | 136 | ||
| GenBank mRNA | BC171580.1 | - | 1 - 2 | 110 | 136 | |
More Information
Set Membership: |
|

srgap1b
Genomic Map