Assay Details
| Gene Symbol: srgap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030095.1 | NP_001025266.1 | 2 - 3 | 734 | 71 | |
| XM_005164583.4 | XP_005164640.1 | 2 - 3 | 752 | 71 | ||
| GenBank mRNA | BC171580.1 | - | 2 - 3 | 305 | 71 | |
More Information
Set Membership: |
|

srgap1b
Genomic Map