Assay Details
| Gene Symbol: ildr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030192.1 | NP_001025363.1 | 7 - 8 | 1130 | 72 | |
| XM_005167845.4 | XP_005167902.1 | 8 - 9 | 1294 | 72 | ||
| XM_005167846.4 | XP_005167903.1 | 8 - 9 | 1291 | 72 | ||
| XM_005167847.4 | XP_005167904.1 | 7 - 8 | 1237 | 72 | ||
| XM_005167848.4 | XP_005167905.1 | 7 - 8 | 1179 | 72 | ||
| XM_005167849.4 | XP_005167906.1 | 7 - 8 | 1176 | 72 | ||
| GenBank mRNA | BC098536.1 | - | 7 - 8 | 1130 | 72 | |
| CU638797.1 | - | 7 - 8 | 1154 | 72 | ||
More Information
Set Membership: |
|

ildr2
Genomic Map