Assay Details
| Gene Symbol: grk7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031841.3 | NP_001027011.2 | 1 - 2 | 722 | 113 | |
| GenBank mRNA | AB212995.1 | - | 1 - 2 | 766 | 113 | |
| BC163587.1 | - | 1 - 2 | 706 | 113 | ||
| EH435535.1 | - | 1 - 2 | 751 | 113 | ||
More Information
Set Membership: |
|

grk7a
Genomic Map