Assay Details
| Gene Symbol: slc5a8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034969.1 | NP_001030141.1 | 6 - 7 | 933 | 68 | |
| GenBank mRNA | AY727859.1 | - | 6 - 7 | 933 | 68 | |
| BC124585.1 | - | 6 - 7 | 840 | 68 | ||
More Information
Set Membership: |
|

slc5a8l
Genomic Map