Assay Details
| Gene Symbol: wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037696.1 | NP_001032785.1 | 5 - 6 | 1099 | 76 | |
| XM_005161848.4 | XP_005161905.1 | 5 - 6 | 1457 | 76 | ||
| GenBank mRNA | BC162350.1 | - | 5 - 6 | 1026 | 76 | |
| BC162832.1 | - | 5 - 6 | 1026 | 76 | ||
| DQ238862.1 | - | 5 - 6 | 1099 | 76 | ||
More Information
Set Membership: |
|

wwtr1
Genomic Map