Assay Details
| Gene Symbol: cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039805.2 | NP_001034894.2 | 4 - 5 | 529 | 71 | |
| GenBank mRNA | AB201385.1 | - | 4 - 5 | 557 | 71 | |
| BC083297.1 | - | 4 - 5 | 571 | 71 | ||
| BC133981.1 | - | 4 - 5 | 529 | 71 | ||
More Information
Set Membership: |
|

cnpy3
Genomic Map