Assay Details
| Gene Symbol: bmpr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039817.1 | NP_001034906.1 | 11 - 12 | 1738 | 74 | |
| GenBank mRNA | DQ002898.1 | - | 11 - 12 | 1738 | 74 | |
More Information
Set Membership: |
|

bmpr2a
Genomic Map