Assay Details
| Gene Symbol: bmpr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039817.1 | NP_001034906.1 | 3 - 4 | 586 | 96 | |
| XM_005165835.4 | XP_005165892.1 | 3 - 4 | 975 | 96 | ||
| GenBank mRNA | DQ002898.1 | - | 3 - 4 | 586 | 96 | |
More Information
Set Membership: |
|

bmpr2a
Genomic Map