Assay Details
| Gene Symbol: rbm24b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328141.1 | NP_001315070.1 | 1 - 2 | 332 | 127 | |
| GenBank mRNA | BC114276.1 | - | 1 - 2 | 333 | 127 | |
More Information
Set Membership: |
|

rbm24b
Genomic Map