Assay Details
| Gene Symbol: bmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040037.1 | NP_001035126.1 | 1 - 2 | 180 | 77 | |
| XM_017357365.2 | XP_017212854.1 | 1 - 2 | 202 | 77 | ||
| GenBank mRNA | AB239771.1 | - | 1 - 2 | 180 | 77 | |
| DQ424857.1 | - | 1 - 2 | 144 | 77 | ||
More Information
Set Membership: |
|

bmp1a
Genomic Map