Assay Details
| Gene Symbol: bmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040037.1 | NP_001035126.1 | 5 - 6 | 729 | 69 | |
| XM_017357365.2 | XP_017212854.1 | 5 - 6 | 751 | 69 | ||
| GenBank mRNA | AB239771.1 | - | 5 - 6 | 729 | 69 | |
| BC163535.1 | - | 5 - 6 | 717 | 69 | ||
More Information
Set Membership: |
|

bmp1a
Genomic Map