Assay Details
| Gene Symbol: rtn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040335.1 | NP_001035425.1 | 1 - 2 | 1249 | 77 | |
| GenBank mRNA | AY555051.1 | - | 1 - 2 | 1249 | 77 | |
| AY555052.1 | - | 2 - 3 | 746 | 77 | ||
| AY555053.1 | - | 1 - 2 | 965 | 77 | ||
| AY555054.1 | - | 1 - 2 | 965 | 77 | ||
| AY555055.1 | - | 1 - 2 | 965 | 77 | ||
| AY555056.1 | - | 1 - 2 | 965 | 77 | ||
| AY555057.1 | - | 1 - 2 | 965 | 77 | ||
| AY555058.1 | - | 1 - 2 | 965 | 77 | ||
| AY555059.1 | - | 1 - 2 | 965 | 77 | ||
| AY555060.1 | - | 1 - 2 | 965 | 77 | ||
| AY555061.1 | - | 1 - 2 | 965 | 77 | ||
| BC068434.1 | - | 1 - 2 | 1111 | 77 | ||
More Information
Set Membership: |
|

rtn4b
Genomic Map