Assay Details
| Gene Symbol: rtn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040335.1 | NP_001035425.1 | 8 - 9 | 2871 | 152 | |
| XM_009307014.2 | XP_009305289.1 | 7 - 8 | 1810 | 152 | ||
| XM_017358799.2 | XP_017214288.1 | 6 - 7 | 618 | 152 | ||
| GenBank mRNA | AY555051.1 | - | 8 - 9 | 2871 | 152 | |
More Information
Set Membership: |
|

rtn4b
Genomic Map