Assay Details
| Gene Symbol: chd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044858.1 | NP_001038323.1 | 10 - 11 | 1770 | 78 | |
| XM_005159352.3 | XP_005159409.1 | 10 - 11 | 1767 | 78 | ||
| XM_021468085.1 | XP_021323760.1 | 10 - 11 | 1770 | 78 | ||
| GenBank mRNA | BC134983.1 | - | 10 - 11 | 1737 | 78 | |
| BC155052.1 | - | 9 - 10 | 1433 | 78 | ||
More Information
Set Membership: |
|

chd4a
Genomic Map