Assay Details
| Gene Symbol: chd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044858.1 | NP_001038323.1 | 37 - 38 | 5630 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chd4a| Gene Symbol: chd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044858.1 | NP_001038323.1 | 37 - 38 | 5630 | 77 | |
Set Membership: |
|