Assay Details
| Gene Symbol: arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044906.2 | NP_001038371.1 | 6 - 7 | 947 | 61 | |
| NM_001351681.2 | NP_001338610.1 | 6 - 7 | 1217 | 61 | ||
| XM_009303441.3 | XP_009301716.1 | 6 - 7 | 1218 | 61 | ||
| XM_009303442.3 | XP_009301717.1 | 6 - 7 | 990 | 61 | ||
| GenBank mRNA | BC117647.1 | - | 6 - 7 | 902 | 61 | |
More Information
Set Membership: |
|

arhgap36
Genomic Map