Assay Details
| Gene Symbol: cacna2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044960.2 | NP_001038425.2 | 22 - 23 | 1966 | 59 | |
| XM_005164746.4 | XP_005164803.1 | 22 - 23 | 2142 | 59 | ||
| XM_009300288.3 | XP_009298563.1 | 22 - 23 | 2144 | 59 | ||
More Information
Set Membership: |
|

cacna2d1a
Genomic Map