Assay Details
| Gene Symbol: cacna2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044960.2 | NP_001038425.2 | 36 - 37 | 3098 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cacna2d1a| Gene Symbol: cacna2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044960.2 | NP_001038425.2 | 36 - 37 | 3098 | 83 | |
Set Membership: |
|