Assay Details
| Gene Symbol: cacna2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044960.2 | NP_001038425.2 | 7 - 8 | 748 | 93 | |
| XM_005164746.4 | XP_005164803.1 | 7 - 8 | 918 | 93 | ||
| XM_009300285.3 | XP_009298560.1 | 7 - 8 | 920 | 93 | ||
| XM_009300286.3 | XP_009298561.1 | 7 - 8 | 920 | 93 | ||
| XM_009300287.3 | XP_009298562.1 | 7 - 8 | 918 | 93 | ||
| XM_009300288.3 | XP_009298563.1 | 7 - 8 | 920 | 93 | ||
More Information
Set Membership: |
|

cacna2d1a
Genomic Map