Assay Details
| Gene Symbol: cdc42bpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045094.1 | NP_001038559.1 | 29 - 30 | 4025 | 65 | |
| XM_005160749.4 | XP_005160806.1 | 31 - 32 | 5821 | 65 | ||
| XM_005160751.4 | XP_005160808.1 | 31 - 32 | 5821 | 65 | ||
| XM_017352506.2 | XP_017207995.1 | 31 - 32 | 5818 | 65 | ||
| XM_017352507.2 | XP_017207996.1 | 31 - 32 | 5818 | 65 | ||
| XM_021468481.1 | XP_021324156.1 | 32 - 33 | 5842 | 65 | ||
| XM_021468482.1 | XP_021324157.1 | 32 - 33 | 5839 | 65 | ||
| XM_021468483.1 | XP_021324158.1 | 32 - 33 | 5842 | 65 | ||
| XM_021468485.1 | XP_021324160.1 | 31 - 32 | 5803 | 65 | ||
| XM_021468486.1 | XP_021324161.1 | 32 - 33 | 5842 | 65 | ||
More Information
Set Membership: |
|

cdc42bpab
Genomic Map