Assay Details
| Gene Symbol: gtf2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045145.1 | NP_001038610.1 | 3 - 4 | 949 | 64 | |
| XM_005167734.4 | XP_005167791.1 | 3 - 4 | 900 | 64 | ||
| XM_017357708.2 | XP_017213197.1 | 3 - 4 | 765 | 64 | ||
More Information
Set Membership: |
|

gtf2e1
Genomic Map