Assay Details
| Gene Symbol: sppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045244.2 | NP_001038709.2 | 12 - 13 | 1336 | 146 | |
| XM_021478911.1 | XP_021334586.1 | 12 - 13 | 1331 | 146 | ||
| GenBank mRNA | AY926457.1 | - | 12 - 13 | 1320 | 146 | |
| BC044512.1 | - | 12 - 13 | 1304 | 146 | ||
| BC116574.1 | - | 12 - 13 | 1321 | 146 | ||
| EH571140.1 | - | 2 - 3 | 326 | 146 | ||
More Information
Set Membership: |
|

sppl2
Genomic Map