Assay Details
| Gene Symbol: sppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045244.2 | NP_001038709.2 | 1 - 2 | 136 | 121 | |
| XM_021478906.1 | XP_021334581.1 | 1 - 2 | 131 | 121 | ||
| XM_021478911.1 | XP_021334586.1 | 1 - 2 | 131 | 121 | ||
| GenBank mRNA | AY926457.1 | - | 1 - 2 | 120 | 121 | |
| BC044512.1 | - | 1 - 2 | 104 | 121 | ||
| BC116574.1 | - | 1 - 2 | 121 | 121 | ||
| EH550022.1 | - | 1 - 2 | 116 | 123 | ||
More Information
Set Membership: |
|

sppl2
Genomic Map