Assay Details
| Gene Symbol: sppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045244.2 | NP_001038709.2 | 2 - 3 | 247 | 84 | |
| XM_021478906.1 | XP_021334581.1 | 2 - 3 | 242 | 84 | ||
| XM_021478911.1 | XP_021334586.1 | 2 - 3 | 242 | 84 | ||
| GenBank mRNA | AY926457.1 | - | 2 - 3 | 231 | 84 | |
| BC044512.1 | - | 2 - 3 | 215 | 84 | ||
| BC116574.1 | - | 2 - 3 | 232 | 84 | ||
| EH550022.1 | - | 2 - 3 | 229 | 84 | ||
More Information
Set Membership: |
|

sppl2
Genomic Map