Assay Details
| Gene Symbol: sppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045244.2 | NP_001038709.2 | 3 - 4 | 434 | 83 | |
| XM_021478906.1 | XP_021334581.1 | 3 - 4 | 429 | 83 | ||
| XM_021478911.1 | XP_021334586.1 | 3 - 4 | 429 | 83 | ||
| GenBank mRNA | AY926457.1 | - | 3 - 4 | 418 | 83 | |
| BC044512.1 | - | 3 - 4 | 402 | 83 | ||
| BC116574.1 | - | 3 - 4 | 419 | 83 | ||
More Information
Set Membership: |
|

sppl2
Genomic Map